Skip to Content
Merck
All Photos(2)

Key Documents

197228

Sigma-Aldrich

BCRP Inhibitor IV, YHO-13351

The BCRP Inhibitor IV, YHO-13351 controls the biological activity of BCRP.

Synonym(s):

BCRP Inhibitor IV, YHO-13351, Diethylaminoacetic acid, 1-(5-(( Z)-2-cyano-2-(3,4-dimethoxyphenyl)vinyl)thiophen-2-yl)piperidin-4-yl ester, Methanesulfonate, Diethylaminoacetic acid, 1-(5-((Z)-2-cyano-2-(3,4-dimethoxyphenyl)vinyl)thiophen-2-yl)piperidin-4-yl ester, Methanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H33N3O4S · xCH4O3S
Molecular Weight:
483.62 (free base basis)
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

yellow

solubility

water: 25 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

O=C(CN(CC)CC)OC(CC1)CCN1C(S2)=CC=C2/C=C(C(C=C3)=CC(OC)=C3OC)\C#N.[xCH3SO3H]

General description

An in vivo bioavailable (~87%; t1/2<5 mins. during p.o. and i.v. administration) and highly water soluble (>100 mg/ml) diethylaminoacetate prodrug of YHO-13177 (Cat. No. 197227) that reverses BCRP-mediated irinotecan resistance in P388/BCRP and HCT116/BCRP xenograft mouse models, and suppresses tumor growth and significantly enhances the survival (100 or 200 mg/kg, i.p.).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Yamazaki, R., et al. 2011. Mol. Cancer Ther.10, 1252.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service