Skip to Content
Merck
All Photos(1)

Key Documents

1.17062

Supelco

SPECTROMELT® C15 Grinding and tableting aid for X-ray fluorescence spectroscopy

Sign Into View Organizational & Contract Pricing


About This Item

Quality Level

Legal Information

SPECTROMELT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service