Skip to Content
Merck
All Photos(1)

Key Documents

1.15926

Sigma-Aldrich

Phloxin B (C.I. 45410)

for microscopy Certistain®

Synonym(s):

Phloxin B (C.I. 45410)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H2Br4Cl4Na2O5
CAS Number:
Molecular Weight:
829.63
MDL number:
UNSPSC Code:
41116130
EC Index Number:
242-355-6

Quality Level

form

solid

potency

2870 mg/kg LD50, oral (Rat)

composition

Dye, ≥80% gravimetric

pH

9.7 (20 °C, 1 g/L in H2O)

solubility

100 g/L

bulk density

650 kg/m3

εmax

930-1400 at 546-550 nm in 50% ethanol at 0.005 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

SMILES string

[Na+].[Na+].Brc1c2c(cc(c1[O-])Br)C4(OC(=O)c5c4c(c(c(c5Cl)Cl)Cl)Cl)c3c(c(c(c(c3)Br)[O-])Br)O2

InChI

1S/C20H4Br4Cl4O5.2Na/c21-5-1-3-17(9(23)15(5)29)32-18-4(2-6(22)16(30)10(18)24)20(3)8-7(19(31)33-20)11(25)13(27)14(28)12(8)26;;/h1-2,29-30H;;/q;2*+1/p-2

InChI key

OOYIOIOOWUGAHD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

The dry dye Phloxin B (C.I. 45410) for microscopy Certistain®, is used for the preparation of a staining solution for cytoplasmic staining in histological sections of human origin. It is mainly used in Hemytoxylin and Eosin (H&E) overview staining that is routinely performed in histological laboratories. Eosin and Phloxine bind to the positively charged plasma proteins. The combination of both dyes results in a broader color range in the tissue. As a Certistain® dye it is chemically quality-certified according to highest standards and strict specifications. As an IVD product and CE registered, it can be used hassle-free in diagnostics and laboratory accreditations. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Dye content (gravimetrically): ≥ 80 %
Identity (UV/VIS-Spectrum): passes test
Absorption maximum λ max (ethanol 50 %): 546 - 550 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.005 g/l; ethanol 50 %): 930 - 1400
TLC-Test: passes test
Loss on drying (110 °C): ≤ 10 %
Suitability for microscopy: passes test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service