Skip to Content
Merck
All Photos(3)

Key Documents

1.13312

Millipore

MULTIRACK

for use with CULTURA mini incubator

Synonym(s):

Holder

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
HUF 33,800.00

HUF 33,800.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 UNIT
HUF 33,800.00

About This Item

UNSPSC Code:
41104424
NACRES:
NB.14

HUF 33,800.00


Please contact Customer Service for Availability

Request more information

packaging

pkg of (1 unit)

application(s)

food and beverages
sample preparation

compatibility

for use with CULTURA mini incubator

storage temp.

15-25°C

General description

The multirack is an accessory that can be used with CULTURA mini incubator. The multirack aids in the orderly placement of the test samples in the mini incubator. The mini incubator is used for the incubation of the samples small in dimension such as petri dishes or contact slides.

Application

Multirack is a CULTURA mini incubator accessory used for the incubation of contact slides and other smaller samples.

Linkage


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service