Skip to Content
Merck
All Photos(1)

Key Documents

1.12513

Supelco

Trifluoroacetic anhydride

for gas chromatography

Synonym(s):

Trifluoroacetic anhydride, Bis(trifluoroacetic) anhydride, TFAA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3CO)2O
CAS Number:
Molecular Weight:
210.03
MDL number:
UNSPSC Code:
12352106
EC Index Number:
206-982-9
NACRES:
NA.21

vapor pressure

405 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

bp

40 °C/1013 hPa

mp

-65 °C

density

1.51 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C4F6O3/c5-3(6,7)1(11)13-2(12)4(8,9)10

InChI key

QAEDZJGFFMLHHQ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service