Skip to Content
Merck
All Photos(1)

Key Documents

1.07049

Supelco

Ethyl methyl ketone

for analysis EMPARTA® ACS

Sign Into View Organizational & Contract Pricing

Select a Size

1070492500
HUF 33,700.00

HUF 33,700.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1070492500
HUF 33,700.00

About This Item

UNSPSC Code:
12191502
grade:
ACS reagent

HUF 33,700.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

ACS reagent

Quality Level

dilution

(for routine analytical applications)

storage temp.

room temp

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

30.2 °F - closed cup

Flash Point(C)

-1 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service