Skip to Content
Merck
All Photos(1)

Key Documents

S-089

Supelco

UR-144 4-Hydroxypentyl metabolite

certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H29NO2
CAS Number:
Molecular Weight:
327.46
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

certified reference material

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

format

single component solution

storage temp.

−20°C

SMILES string

O=C(C1C(C)(C)C1(C)C)C2=CN(CCCC(O)C)C3=CC=CC=C32

InChI

1S/C21H29NO2/c1-14(23)9-8-12-22-13-16(15-10-6-7-11-17(15)22)18(24)19-20(2,3)21(19,4)5/h6-7,10-11,13-14,19,23H,8-9,12H2,1-5H3

InChI key

YHFHYHZNLCEAAP-UHFFFAOYSA-N

General description

A certified solution standard suitable for use as starting material in calibrators and controls for GC/MS or LC/MS synthetic cannabinoid testing methods in forensic analysis, clinical toxicology, or urine drug testing. UR-144 5-Hydroxypentyl metabolite is a primary urinary metabolite of UR-144, a third generation synthetic cannabinoid that has seen wide popularity in illicit drug markets since late 2012. This biomarker is also referred to as the terminal omega hydroxyl metabolite of UR-144.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service