Skip to Content
Merck
All Photos(1)

Key Documents

ATE341034090

AstaTech, Inc.

N-ISOPROPYL-2-OXOOXAZOLIDINE-3-SULFONAMIDE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
208.24
MDL number:
Pricing and availability is not currently available.

Iupac Name

N-isopropyl-2-oxo-1,3-oxazolidine-3-sulfonamide

Inchi Code

1S/C6H12N2O4S/c1-5(2)7-13(10,11)8-3-4-12-6(8)9/h5,7H,3-4H2,1-2H3

InChI key

LWTSNAAKXGLFDJ-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

95%

Shipping Temperature

Normal

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=10725

Signal Word

Warning

Hazard Statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=AR3692


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library