Key Documents
Z743199
Julabo® bath test tube racks
to fit 90 test tubes, diam. 12 mm (13 mm), stainless steel
Synonym(s):
tube holder
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
Pricing and availability is not currently available.
material
stainless steel
packaging
pack of 1 ea
manufacturer/tradename
Julabo 8970345
parameter
100 °C max. temp.
diam.
12 mm (13 mm)
General description
Stainless steel rack suitable for temperatures up to 100°C. If solvent based samples or acidic samples are tempered above 60°C, it is recommended using test glass inserts made of stainless steel.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)