Skip to Content
Merck
All Photos(1)

Key Documents

PH006805

Sigma-Aldrich

1-(4-Bromo-2-hydroxyphenyl)-1-propanone

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C9H9BrO2
CAS Number:

Pricing and availability is not currently available.

SMILES string

OC1=C(C=CC(Br)=C1)C(CC)=O

InChI

1S/C9H9BrO2/c1-2-8(11)7-4-3-6(10)5-9(7)12/h3-5,12H,2H2,1H3

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
359912ALD00438ALD00434
reaction suitability

reaction type: C-C Bond Formation

reaction suitability

reaction type: C-C Bond Formation

reaction suitability

reaction type: C-C Bond Formation

reaction suitability

reaction type: C-C Bond Formation

form

powder

form

powder

form

powder

form

powder

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service