Skip to Content
Merck
All Photos(2)

Key Documents

C107

Sigma-Aldrich

Calcium acetylacetonate hydrate

99.95%

Synonym(s):

2,4-Pentanedione calcium derivative, Ca(acac)2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca(C5H7O2)2 · xH2O
CAS Number:
Molecular Weight:
238.29 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

99.95%

form

solid

composition

Degree of hydration, ~0.5

reaction suitability

core: calcium

mp

>280 °C (dec.) (lit.)

SMILES string

O.CC(=O)\C=C(\C)O[Ca]O\C(C)=C/C(C)=O

InChI

1S/2C5H8O2.Ca.H2O/c2*1-4(6)3-5(2)7;;/h2*3,6H,1-2H3;;1H2/q;;+2;/p-2/b2*4-3-;;

InChI key

FFWWBMKQRYMOKA-SUKNRPLKSA-L

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service