Skip to Content
Merck
All Photos(1)

Key Documents

901349

Sigma-Aldrich

Hexagonal boron nitride ink

greener alternative

for blade coating

Synonym(s):

Hexagonal boron nitride dispersion, with ethyl lactate, hBN blade coatable ink, hBN dispersion

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BN
CAS Number:
Molecular Weight:
24.82
MDL number:
UNSPSC Code:
12352302

form

dispersion

composition

Solid content, 20-40%

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

white

particle size

≤0.3 μm

surface tension

28-32 dyn/cm

viscosity

200-450 cP (shear viscosity at 1000 s-1, 25 °C)

bp

152-156 °C (ethyl lactate)

density

0.9-1.1 g/mL at 25 °C

greener alternative category

InChI

1S/BN/c1-2

InChI key

PZNSFCLAULLKQX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Curing condition: 250-350 °C, 20-30 min
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". Boron nitride materials have interesting properties like large energy band gap, good resistance to oxidation, excellent thermal stability, thermal conductivity, chemical inertness, significance mechanical property and widespred applications. Click here for more information.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

116.6 °F

Flash Point(C)

47 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Recent advancements in paper-based sensing platforms offer cost-effective clinical diagnostics with microfluidic channels and colorimetric or electrochemical detection zones.

Carbon-based Sustainable Organic Electronics (SOE) limit the use of critical elements and biodegrade at their end-of-life. This review offers insight on how structural and energy disorder in these materials influence device performance and includes evaluations of various transport models and their limitations.

Review on 1D vdWHs: Discusses materials, synthesis, optoelectronic applications, challenges, and future perspectives for 1D vdWH-based devices.

Professors summarize recent 2D materials synthesis advancements and biosensing applications in various fields.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service