Skip to Content
Merck
All Photos(1)

Key Documents

900800

Sigma-Aldrich

ITIC-Th

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C86H74N4O2S8
CAS Number:
Molecular Weight:
1452.05
UNSPSC Code:
12352101
To inquire about this product 900800, please contact your local Merck office or dealer. Contact Technical Service

description

Band gap: 1.73 eV

Assay

99%

form

solid

solubility

chlorobenzene: soluble
chloroform: soluble
dichlorobenzene: soluble

Orbital energy

HOMO -5.66 eV 
LUMO -3.93 eV 

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly Efficient Parallel-?Like Ternary Organic Solar Cells.
Tao L, et al.
Chemistry of Materials, 29(7), 2914-2920 (2017)
Yuze Lin et al.
Journal of the American Chemical Society, 138(14), 4955-4961 (2016-03-26)
We develop an efficient fused-ring electron acceptor (ITIC-Th) based on indacenodithieno[3,2-b]thiophene core and thienyl side-chains for organic solar cells (OSCs). Relative to its counterpart with phenyl side-chains (ITIC), ITIC-Th shows lower energy levels (ITIC-Th: HOMO = -5.66 eV, LUMO =

Articles

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service