Skip to Content
Merck
All Photos(2)

Key Documents

772852

Sigma-Aldrich

Yttrium borate

powder, -200 mesh, 99.9% trace rare earth metals basis

Synonym(s):

Yttrium orthoborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
YBO3
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace rare earth metals basis

form

powder

particle size

-200 mesh

mp

>300 °C

SMILES string

[Y+3].[O-]B([O-])[O-]

InChI

1S/BO3.Y/c2-1(3)4;/q-3;+3

InChI key

FERAQKHYRHQYKD-UHFFFAOYSA-N

Application

YBO3 is a promising phosphor host material, showing different emission properties upon doping with rare earth and other elements. The borate based materials find applications in solid state lighting, LED devices and also as photon conversion material.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chawla, S.
Applied Surface Science, 257, 7167-7167 (2011)
Blasse, G.; et al.
J. Chem. Phys. , 47, 5139-5139 (1967)
Wei, Z-G.
Chemistry of Materials, 15, 3011-3011 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service