Skip to Content
Merck
All Photos(1)

Key Documents

757519

Sigma-Aldrich

3-Chloro-D-alanine methyl ester hydrochloride

97%

Synonym(s):

β-Chloro-D-alanine methyl ester hydrochloride, (S)-Methyl 2-amino-3-chloropropanoate hydrochloride, (S)-Methyl 2-amino-3-chloropropionate hydrochloride, 3-Chloro-D-Ala-OMe hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8ClNO2·HCl
CAS Number:
Molecular Weight:
174.03
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

optical activity

[α]22/D +4.5°, c = 0.5% in DMSO

mp

143-148 °C

application(s)

peptide synthesis

functional group

amine
chloro
ester

storage temp.

2-8°C

SMILES string

Cl.COC(=O)[C@H](N)CCl

InChI

1S/C4H8ClNO2.ClH/c1-8-4(7)3(6)2-5;/h3H,2,6H2,1H3;1H/t3-;/m1./s1

InChI key

POPBCSXDEXRDSX-AENDTGMFSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Damjan Makuc et al.
Molecules (Basel, Switzerland), 25(7) (2020-04-11)
d-cycloserine is a broad-spectrum antibiotic that is currently being used as a secondary choice in the treatment of tuberculosis. In recent years, it has become more popular, due to its effect on the nervous system. In this current study, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service