Skip to Content
Merck
All Photos(1)

Key Documents

731072

Sigma-Aldrich

2-Bromo-5-fluoro-3-methylpyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5BrFN
CAS Number:
Molecular Weight:
190.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.540

density

1.598 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

Cc1cc(F)cnc1Br

InChI

1S/C6H5BrFN/c1-4-2-5(8)3-9-6(4)7/h2-3H,1H3

InChI key

QLBXFAJGMGFVCN-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

206.0 °F

Flash Point(C)

96.67 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service