Skip to Content
Merck
All Photos(1)

Key Documents

707708

Sigma-Aldrich

5-Hexen-2-yn-1-ol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O
CAS Number:
Molecular Weight:
96.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

liquid

refractive index

n20/D 1.478

density

0.926 g/mL at 25 °C

SMILES string

OCC#CCC=C

InChI

1S/C6H8O/c1-2-3-4-5-6-7/h2,7H,1,3,6H2

InChI key

KSOMTIQGBYHOJM-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

170.1 °F

Flash Point(C)

76.7 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service