Skip to Content
Merck
All Photos(2)

Key Documents

697508

Sigma-Aldrich

Hafnium isopropoxide isopropanol adduct

99.9% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H28HfO4
CAS Number:
Molecular Weight:
414.84
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

99.9% trace metals basis

form

crystalline powder
crystals

reaction suitability

core: hafnium

impurities

~1% Zr

SMILES string

CC(C)O[Hf](OC(C)C)(OC(C)C)OC(C)C

InChI

1S/4C3H7O.Hf/c4*1-3(2)4;/h4*3H,1-2H3;/q4*-1;+4

InChI key

HRDRRWUDXWRQTB-UHFFFAOYSA-N

Application

Hafnium isopropoxide isopropanol adduct was used as a precursor to prepare mesoporous HfO2 by the thermohydrolytic approach. The high active surface area and uniform porosity enables these HfO2 to be used as adsorbents for sequestration of heavy metal ions.[1]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

General Facile Approach to Transition-Metal Oxides with Highly Uniform Mesoporosity and Their Application as Adsorbents for Heavy-Metal-Ion Sequestration.
Seisenbaeva GA, et al.
Chemistry?A European Journal , 20(34), 10732-10736 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service