Skip to Content
Merck
All Photos(1)

Key Documents

697214

Sigma-Aldrich

1,3-Dichloro-6-fluoroisoquinoline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H4Cl2FN
CAS Number:
Molecular Weight:
216.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

needles

mp

131-133 °C

functional group

chloro
fluoro

SMILES string

Fc1ccc2c(Cl)nc(Cl)cc2c1

InChI

1S/C9H4Cl2FN/c10-8-4-5-3-6(12)1-2-7(5)9(11)13-8/h1-4H

InChI key

BDAYBRFYRZTMGQ-UHFFFAOYSA-N

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service