Skip to Content
Merck
All Photos(1)

Key Documents

689319

Sigma-Aldrich

Dehydroacetic acid

Lonza quality, 99.0-100.3 % (w/w) (T)

Synonym(s):

2-Acetyl-5-hydroxy-3-oxo-4-hexenoic acid δ-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8O4
CAS Number:
Molecular Weight:
168.15
Beilstein:
6129
EC Number:
MDL number:
UNSPSC Code:
12352100
Pricing and availability is not currently available.

grade

Lonza quality

form

crystalline
powder

manufacturer/tradename

Lonza Ltd

concentration

99.0-100.3 % (w/w) (T)

bp

270 °C (lit.)

mp

109.0-111.0 °C (opt. transm.)
111-113 °C (lit.)

SMILES string

CC(=O)C1C(=O)OC(C)=CC1=O

InChI

1S/C8H8O4/c1-4-3-6(10)7(5(2)9)8(11)12-4/h3,7H,1-2H3

InChI key

PGRHXDWITVMQBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

transparence: >63.2%; 420 nm, 10 mm 1g/30 mL MeOH

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

314.6 °F

Flash Point(C)

157 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service