Skip to Content
Merck
All Photos(1)

Key Documents

685690

Sigma-Aldrich

Poly[(9,9-dihexylfluoren-2,7-diyl)-alt-(2,5-dimethyl-1,4-phenylene)]

Synonym(s):

PF-DMB

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

powder

mp

>400 °C

fluorescence

λex 330 nm; λem 379 nm in chloroform

Features and Benefits

Polymer end-capped with 3,5-dimethylbenzene.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service