Skip to Content
Merck
All Photos(1)

Key Documents

672459

Sigma-Aldrich

2-(Dicyclohexylphosphino)-1-(2-methoxyphenyl)-1H-pyrrole

95%

Synonym(s):

cataCXium® POMeCy

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H32NOP
CAS Number:
Molecular Weight:
369.48
Beilstein:
9656639
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

95%

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
reaction type: Asymmetric synthesis

functional group

phosphine

SMILES string

COc1ccccc1-n2cccc2P(C3CCCCC3)C4CCCCC4

InChI

1S/C23H32NOP/c1-25-22-16-9-8-15-21(22)24-18-10-17-23(24)26(19-11-4-2-5-12-19)20-13-6-3-7-14-20/h8-10,15-20H,2-7,11-14H2,1H3

InChI key

JUZAKZRALSJLOV-UHFFFAOYSA-N

General description

sold in collaboration with Solvias AG

Application

  • Used as ligand in arylation reaction with bromobenzene, and phosphorylation reactions with chlorophosphines
  • Cocatalyst in palladium-catalyzed Suzuki coupling reactions

Legal Information

WO 2004/101581
cataCXium is a registered trademark of Umicore AG & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service