Skip to Content
Merck
All Photos(3)

Key Documents

637181

Sigma-Aldrich

4-(tert-Butyldimethylsiloxy)phenol

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
HUF 56,200.00
5 G
HUF 168,600.00

HUF 56,200.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
HUF 56,200.00
5 G
HUF 168,600.00

About This Item

Empirical Formula (Hill Notation):
C12H20O2Si
CAS Number:
Molecular Weight:
224.37
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

HUF 56,200.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

60-64 °C (lit.)

SMILES string

CC(C)(C)[Si](C)(C)Oc1ccc(O)cc1

InChI

1S/C12H20O2Si/c1-12(2,3)15(4,5)14-11-8-6-10(13)7-9-11/h6-9,13H,1-5H3

InChI key

KZTVHIZALLBXMO-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service