Skip to Content
Merck
All Photos(1)

Key Documents

634735

Sigma-Aldrich

Lanthanum aluminum oxide

single crystal substrate, <100>, ≥99.99% trace metals basis

Synonym(s):

Lanthanum aluminate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LaAlO3
CAS Number:
Molecular Weight:
213.89
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

description

single side polished

Assay

≥99.99% trace metals basis

dielectric constant

~25

reaction suitability

reagent type: catalyst
core: lanthanum

size

10 mm × 10 mm × 0.5 mm

mp

2080 °C (lit.)

density

6.52 g/mL at 25 °C (lit.)

semiconductor properties

<100>

SMILES string

O=[Al]O[La]=O

InChI

1S/Al.La.3O

InChI key

KJXBRHIPHIVJCS-UHFFFAOYSA-N

Features and Benefits

Single crystal lanthanum aluminate substrate lattices have a good lattice match with most perovskite-structured materials. It is an excellent substrate for the epitaxial growth of magnetic and ferro-electric thin films and high-temperature superconductors. Due to its dielectric properties it is also posses properties for low-loss microwave and dielectric resonance applications.

Physical form

Rhombohedral at 25 °C (a = 5.357 Å, c = 13.22 Å); Cubic at >435 °C (a = 3.821 Å)

Other Notes

thermal expansion = 10 × 10-6/°C; loss tangent at 10 GHz = ~3 × 10-4 @ 300 K, 0.6 × 10-4 @ 77 K; insoluble in mineral acids at 25 °C, soluble in H3PO3 at >150 °C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service