Skip to Content
Merck
All Photos(1)

Key Documents

59900

Sigma-Aldrich

Methyl isovalerate

≥98.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2COOCH3
CAS Number:
Molecular Weight:
116.16
Beilstein:
1699922
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (GC)

refractive index

n20/D 1.393 (lit.)
n20/D 1.393

bp

114-115 °C (lit.)

density

0.881 g/mL at 25 °C (lit.)

SMILES string

COC(=O)CC(C)C

InChI

1S/C6H12O2/c1-5(2)4-6(7)8-3/h5H,4H2,1-3H3

InChI key

OQAGVSWESNCJJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

57.9 °F - closed cup

Flash Point(C)

14.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jechan Lee et al.
Environmental pollution (Barking, Essex : 1987), 230, 81-86 (2017-06-27)
This study lays great emphasis on establishing a reliable analytical platform to quantify and specify volatile fatty acids (VFAs) in the aqueous phase by derivatizing VFAs into their corresponding alkyl esters via thermally-induced rapid esterification (only 10 s reaction time). To
Sang-Ryong Lee et al.
The Science of the total environment, 610-611, 992-996 (2017-08-26)
This report proposes a new approach to evaluate the odour nuisance of cattle manure samples from three different cattle breeds (i.e., native cattle, beef cattle, and milk cow) by means of quantification and speciation of volatile fatty acids (VFAs). To

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service