Skip to Content
Merck
All Photos(1)

Documents

51462

Sigma-Aldrich

Urea phosphate salt

≥98.0% (N)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CONH2 · H3PO4
CAS Number:
Molecular Weight:
158.05
Beilstein:
3709707
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥98.0% (N)

form

solid

mp

116-118 °C (dec.) (lit.)

solubility

H2O: 50 mg/mL, clear

SMILES string

NC(N)=O.OP(O)(O)=O

InChI

1S/CH4N2O.H3O4P/c2-1(3)4;1-5(2,3)4/h(H4,2,3,4);(H3,1,2,3,4)

InChI key

DZHMRSPXDUUJER-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonia Ressler et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 108(4), 1697-1709 (2019-11-19)
Biomimetic triphasic strontium-substituted calcium phosphate (CaP) powders were prepared by wet precipitation method at 50°C, using CaCO3 , (NH2 )2 COH3 PO4 , and Sr(NO3 )2 as reagents. Calcite was prepared from biogenic source (cuttlefish bone). The synthesized powders have
Antonia Ressler et al.
Carbohydrate polymers, 197, 469-477 (2018-07-17)
Injectable hydrogels have emerged as promising biomaterials for tissue engineering applications. The goal of this study was to evaluate the potential of a pH-responsive chitosan-hydroxyapatite hydrogel to be used as a three-dimensional support for encapsulated mesenchymal stem cells (MSCs) osteogenic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service