Skip to Content
Merck
All Photos(3)

Key Documents

489689

Sigma-Aldrich

3-Methyl-5-isoxazoleacetic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7NO3
CAS Number:
Molecular Weight:
141.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

101-104 °C (lit.)

functional group

carboxylic acid

SMILES string

Cc1cc(CC(O)=O)on1

InChI

1S/C6H7NO3/c1-4-2-5(10-7-4)3-6(8)9/h2H,3H2,1H3,(H,8,9)

InChI key

POEFJFLAFQWOTL-UHFFFAOYSA-N

General description

3-Methyl-5-isoxazoleacetic acid is a substituted isoxazole.

Application

3-Methyl-5-isoxazoleacetic acid may be used as a reagent in the solid phase synthesis of von Hippel-Lindau protein (VHL) ligands.[1] It may also be used to prepare (N-(4-chlorophenyl)-α-[[(4-chlorophenyl)amino]methylene]-3-methyl-5-isoxazoleacetamide).[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timothy B Johnstone et al.
The Journal of pharmacology and experimental therapeutics, 336(3), 908-915 (2010-12-17)
α5 Subunit-containing GABA(A) receptors (GABA(A)Rs) and α7 neuronal nicotinic-acetylcholine receptors (nAChRs) are members of the Cys-loop family of ligand-gated ion channels (LGICs) that mediate cognitive and attentional processes in the hippocampus. α5 GABA(A)Rs alter network activity by tonic inhibition of
Dennis L Buckley et al.
Journal of the American Chemical Society, 134(10), 4465-4468 (2012-03-01)
E3 ubiquitin ligases, which bind protein targets, leading to their ubiquitination and subsequent degradation, are attractive drug targets due to their exquisite substrate specificity. However, the development of small-molecule inhibitors has proven extraordinarily challenging as modulation of E3 ligase activities

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service