Skip to Content
Merck
All Photos(1)

Key Documents

483249

Sigma-Aldrich

Antimony(V) oxide

85%

Synonym(s):

Antimony pentoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sb2O5
CAS Number:
Molecular Weight:
323.52
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

85%

mp

380 °C (dec.) (lit.)

density

3.78 g/mL at 25 °C (lit.)

SMILES string

O=[Sb](=O)O[Sb](=O)=O

InChI

1S/5O.2Sb

InChI key

LJCFOYOSGPHIOO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service