Skip to Content
Merck
All Photos(1)

Key Documents

454966

Sigma-Aldrich

mono-2-(Acryloyloxy)ethyl succinate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCO2CH2CH2O2CCH2CH2CO2H
CAS Number:
Molecular Weight:
216.19
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
Pricing and availability is not currently available.

contains

1000 ppm BHT as inhibitor
250 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.465 (lit.)

density

1.23 g/mL at 25 °C (lit.)

SMILES string

OC(=O)CCC(=O)OCCOC(=O)C=C

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service