Skip to Content
Merck
All Photos(3)

Key Documents

342211

Sigma-Aldrich

Nickel(II) fluoride

Synonym(s):

Nickel difluoride, Nickel fluoride, Nickelous fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiF2
CAS Number:
Molecular Weight:
96.69
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder
solubility:
diethyl ether: insoluble(lit.)
ethanol: insoluble(lit.)
To inquire about this product 342211, please contact your local Merck office or dealer. Contact Technical Service

form

powder

reaction suitability

reagent type: catalyst
core: nickel

mp

1380 °C (lit.)

solubility

diethyl ether: insoluble(lit.)
ethanol: insoluble(lit.)

density

4.72 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

F[Ni]F

InChI

1S/2FH.Ni/h2*1H;/q;;+2/p-2

InChI key

DBJLJFTWODWSOF-UHFFFAOYSA-L

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Eye Dam. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service