Skip to Content
Merck
All Photos(1)

Key Documents

341126

Sigma-Aldrich

(Bicyclo[2.2.1]hepta-2,5-diene)[1,4-bis(diphenylphosphino)butane]rhodium(I) tetrafluoroborate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
HUF 38,430.00

HUF 38,430.00

List PriceHUF 54,900.00Save 30%

Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
HUF 38,430.00

About This Item

Empirical Formula (Hill Notation):
C35H36BF4P2Rh
CAS Number:
Molecular Weight:
708.32
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

HUF 38,430.00

List PriceHUF 54,900.00Save 30%

Please contact Customer Service for Availability

Request a Bulk Order

reaction suitability

core: rhodium
reagent type: catalyst

mp

210 °C (dec.) (lit.)

SMILES string

[Rh+].F[B-](F)(F)F.C1C2C=CC1C=C2.C(CCP(c3ccccc3)c4ccccc4)CP(c5ccccc5)c6ccccc6

InChI

1S/C28H28P2.C7H8.BF4.Rh/c1-5-15-25(16-6-1)29(26-17-7-2-8-18-26)23-13-14-24-30(27-19-9-3-10-20-27)28-21-11-4-12-22-28;1-2-7-4-3-6(1)5-7;2-1(3,4)5;/h1-12,15-22H,13-14,23-24H2;1-4,6-7H,5H2;;/q;;-1;+1

InChI key

FVKIJNAEINIHMG-UHFFFAOYSA-N

Other Notes

Contains up to 1 mole acetone of crystallization

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Baptiste Joalland et al.
Angewandte Chemie (International ed. in English), 59(22), 8654-8660 (2020-03-25)
Radio amplification by stimulated emission of radiation (RASER) was recently discovered in a low-field NMR spectrometer incorporating a highly specialized radio-frequency resonator, where a high degree of proton-spin polarization was achieved by reversible parahydrogen exchange. RASER activity, which results from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service