Skip to Content
Merck
All Photos(1)

Key Documents

296899

Sigma-Aldrich

4-Nitrobenzenesulfenyl chloride

technical grade, 95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4SCl
CAS Number:
Molecular Weight:
189.62
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

95%

bp

125 °C/0.1 mmHg (lit.)

mp

48-51 °C (lit.)

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc(SCl)cc1

InChI

1S/C6H4ClNO2S/c7-11-6-3-1-5(2-4-6)8(9)10/h1-4H

InChI key

NCBOVAWEMBIIFK-UHFFFAOYSA-N

General description

4-Nitrobenzenesulfenyl chloride has been used in n-Bu4NI-induced electrophilic cyclization of N,N-dimethyl-(2-phenylethynyl) aniline[1].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Chen et al.
Organic letters, 11(1), 173-176 (2008-12-03)
3-Sulfenyl- and 3-selenylindoles are prepared in excellent yields by the palladium/copper-catalyzed crossing coupling of N,N-dialkyl-o-iodoanilines and terminal alkynes, followed by electrophilic cyclization with arylsulfenyl chlorides and arylselenyl chlorides in the presence of a stoichiometric amount of n-Bu(4)NI.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service