Skip to Content
Merck
All Photos(1)

Key Documents

261602

Sigma-Aldrich

trans-4-Pentylcyclohexanecarboxylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4C6H10CO2H
CAS Number:
Molecular Weight:
198.30
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

51-53 °C (lit.)

functional group

carboxylic acid

SMILES string

CCCCC[C@@H]1CC[C@H](CC1)C(O)=O

InChI

1S/C12H22O2/c1-2-3-4-5-10-6-8-11(9-7-10)12(13)14/h10-11H,2-9H2,1H3,(H,13,14)/t10-,11-

InChI key

RVLAXPQGTRTHEV-XYPYZODXSA-N

Application

trans-4-Pentylcyclohexanecarboxylic acid has been used in preparation of:
  • 4-[(2-(trans-4-pentylcyclohexyl)ethyl]phenol[1]
  • 4-[(4-trans-pentyl-cyclohexanecarbonyl)-(4-pyridin-4-yl-benzyl)-amino]-piperidine-1-tert-butyl carbamate[2]
  • 2-cyano-1-benzofuran-5-yl trans-4-pentylcyclohexanecarboxylate[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Comparative studies of three-and four-ring mesogenic esters containing p-carborane, bicyclo [2.2. 2] octane, cyclohexane, and benzene
Ringstrand B, et al.
Liq. Cryst., 32(8), 1061-1070 (2005)
Synthesis of plasmepsin II inhibitors-potential antimalarial agents
Mueller R, et al.
Molecules (Basel), 8(7), 556-564 (2003)
The synthesis and transition temperatures of 2-(4-alkyl-and 4-alkoxy-phenyl)-5-cyano-1-benzofurans and related diaryl-1-benzofurans-an assessment of how deviations from linearity and conformational effects in a core unit affect mesogenicity.
Friedman MR, et al.
Journal of Materials Chemistry, 11(11), 2759-2772 (2001)
Beili Wang et al.
Environmental science & technology, 53(2), 1022-1030 (2018-12-18)
The nontargeted scanning chemical profiling approach has shown great potential to identify unknown pollutants or novel biological markers; however, the structure identification of unknown compounds is a challenge. In this study, a carboxyl-specific derivatization reagent, N-(4-aminomethylphenyl) pyridinium (AMPP), was coupled

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service