Skip to Content
Merck
All Photos(1)

Key Documents

260401

Sigma-Aldrich

3-Methoxy-1,2-propanediol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
106.12
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.444 (lit.)

bp

220 °C (lit.)

density

1.114 g/mL at 25 °C (lit.)

SMILES string

COCC(O)CO

InChI

1S/C4H10O3/c1-7-3-4(6)2-5/h4-6H,2-3H2,1H3

InChI key

PSJBSUHYCGQTHZ-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Schuff-Werner et al.
Cryobiology, 25(6), 487-494 (1988-12-01)
1-O-methyl-rac-glycerol (1-O-MG), also known as 3-methoxy-1,2-propanediol is a lipophilic derivative of glycerol, and has been studied as a new cryoprotective agent (CPA) for mononuclear blood cells (MNC), a well-established experimental model in cryopreservation. The chemical modification of the glycerol molecule
Carsten Fauhl et al.
Journal of AOAC International, 87(5), 1179-1188 (2004-10-21)
The aim of the present study was to provide the official wine control authorities with an internationally validated method for the determination of 3-methoxy-1,2-propanediol (3-MPD) and cyclic diglycerols (CycDs)-both of which are recognized as impurities of technical glycerol-in different types
Neepa Choksi et al.
Cutaneous and ocular toxicology, 39(3), 180-192 (2020-06-27)
OptiSafe is an in chemico test method that identifies potential eye irritants based on macromolecular damage following test chemical exposure. The OptiSafe protocol includes a prescreen assessment that identifies test chemicals that are outside the applicability domain of the test

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service