Skip to Content
Merck
All Photos(1)

Key Documents

243906

Sigma-Aldrich

Borane triphenylphosphine complex

97%

Synonym(s):

Triphenylphosphineborane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3P · BH3
CAS Number:
Molecular Weight:
276.12
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reagent type: reductant

mp

189-191 °C (lit.)

SMILES string

B.c1ccc(cc1)P(c2ccccc2)c3ccccc3

InChI

1S/C18H15P.BH3/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;/h1-15H;1H3

InChI key

CHVBILDTVBDARQ-UHFFFAOYSA-N

Application

Reactant for:
  • C-P cross coupling with aryl iodides
  • Synthesis of N-heterocyclic carbene borane complexes via Lewis base exchange
  • Catalyst-free alcoholysis to deprotect phosphane-boranes
  • Undergoing heterolytic activation
  • Deprotection via reactions with resin-supported amines
  • Insertion of overcrowded silylene into hydroboranes

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service