Skip to Content
Merck
All Photos(1)

Key Documents

178969

Sigma-Aldrich

Tri-sec-butylborane solution

1.0 M in THF

Synonym(s):

Tris(1-methylpropyl)borane

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
CHF 255.69
1 G
CHF 495.32

Vendor SKU: AC2005-250MG

CHF 255.69


Availability
Ships in 14 days - from Aldrich Partner.

Request a Bulk Order

Select a Size

Change View
250 MG
CHF 255.69
1 G
CHF 495.32

About This Item

Linear Formula:
[C2H5CH(CH3)]3B
CAS Number:
Molecular Weight:
182.15
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Vendor SKU: AC2005-250MG

CHF 255.69


Availability
Ships in 14 days - from Aldrich Partner.

Request a Bulk Order

Quality Level

reaction suitability

reagent type: reductant

concentration

1.0 M in THF

density

0.799 g/mL at 25 °C

SMILES string

CCC(C)B(C(C)CC)C(C)CC

InChI

1S/C12H27B/c1-7-10(4)13(11(5)8-2)12(6)9-3/h10-12H,7-9H2,1-6H3

InChI key

YUPAWYWJNZDARM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
  • Allylic alkylations for synthesis of unsaturated aldehydes and alcohols
  • Cis-addition of carbon-hydrogen bonds of pyrroles and thiophenes to alkynes
  • Borane-induced radical reduction of alkenyl and alkynyliodanes
  • Initiating polymerization for synthesis of polymethylenes and ethylene-propylene
  • Asymmetric synthesis of vinylglycidols
  • Asymmetric addition of alcohols to vinyl epoxides

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service