Skip to Content
Merck
All Photos(2)

Key Documents

151408

Sigma-Aldrich

4-Methyl-3-nitrobenzoic acid

99%

Synonym(s):

3-Nitro-p-toluic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)CO2H
CAS Number:
Molecular Weight:
181.15
Beilstein:
1874411
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

187-190 °C (lit.)

functional group

carboxylic acid
nitro

SMILES string

Cc1ccc(cc1[N+]([O-])=O)C(O)=O

InChI

1S/C8H7NO4/c1-5-2-3-6(8(10)11)4-7(5)9(12)13/h2-4H,1H3,(H,10,11)

InChI key

BBEWSMNRCUXQRF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Methyl-3-nitrobenzoic acid was used in the synthesis of one-dimensional (1D) lanthanide coordination complexes[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jumei Tian et al.
Dalton transactions (Cambridge, England : 2003), 42(23), 8504-8511 (2013-05-01)
Three novel one-dimensional (1D) lanthanide coordination complexes involving the 4-methyl-3-nitrobenzoic acid (HL) ligand, with the general formulae [Gd(L)3(H2O)(CH3OH)] (1), [Gd(L)3(H2O)2]·(4,4'-bpy)·CH3OH (2), [Dy(L)3(H2O)(CH3OH)] (3), have been synthesized by solvothermal reactions. These complexes have been structurally characterized by single-crystal X-ray diffraction, IR

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service