Skip to Content
Merck
All Photos(1)

Key Documents

103977

Sigma-Aldrich

2,3,4,5,6-Pentafluorotoluene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6F5CH3
CAS Number:
Molecular Weight:
182.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

Assay

99%

refractive index

n20/D 1.403 (lit.)

bp

117 °C (lit.)

mp

−30 °C (lit.)

density

1.439 g/mL at 25 °C (lit.)

SMILES string

Cc1c(F)c(F)c(F)c(F)c1F

InChI

1S/C7H3F5/c1-2-3(8)5(10)7(12)6(11)4(2)9/h1H3

InChI key

SXPRVMIZFRCAGC-UHFFFAOYSA-N

General description

2,3,4,5,6-Pentafluorotoluene is obtained by the Friedel Craft′s alkylation of pentafluorobenzene with methyl chloride[1].

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Friedel-Crafts alkylation of pentafluorobenzene.
Beckert WF and Lowe Jr JU.
The Journal of Organic Chemistry, 32(4), 1212-1215 (1967)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service