Merck
HPLC Analysis of FMOC-Serine Enantiomers on Astec® (R,R) P-CAP
HPLC Analysis of FMOC-Serine Enantiomers on Astec® (R,R) P-CAP™ application for HPLC
材料
標準品
产品编号
说明
价格
L-丝氨酸
ReagentPlus®, ≥99% (HPLC)
L-丝氨酸
from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
DL-丝氨酸
BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
DL-丝氨酸
≥98% (TLC)
D -丝氨酸
≥98% (TLC)
L-丝氨酸
BioUltra, ≥99.5% (NT)
CONDITIONS
column
Astec (R,R) P-CAP, 25 cm x 4.6 mm I.D., 5 μm particles (31024AST)
mobile phase
20 mM ammonium acetate in: (A) acetonitrile; (B) methanol; (70:30, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
0.5 μL
sample
1 mg/mL in methanol
说明
法律資訊
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
P-CAP is a trademark of Sigma-Aldrich Co. LLC