跳转至内容
Merck
  • Vitamins and iron blood biomarkers are associated with blood pressure levels in European adolescents. The HELENA study.
Vitamins and iron blood biomarkers are associated with blood pressure levels in European adolescents. The HELENA study.
Nutrition (Burbank, Los Angeles County, Calif.) (2014-09-10)
Augusto César Ferreira de Moraes, Luis Gracia-Marco, Iris Iglesia, Marcela González-Gross, Christina Breidenassel, Marika Ferrari, Dénes Molnar, Sonia Gómez-Martínez, Odysseas Androutsos, Anthony Kafatos, Magdalena Cuenca-García, Michael Sjöström, Frederic Gottrand, Kurt Widhalm, Heráclito Barbosa Carvalho, Luis A Moreno
摘要
Previous research showed that low concentration of biomarkers in the blood during adolescence (i.e., iron status; retinol; and vitamins B6, B12, C, and D) may be involved in the early stages of development of many chronic diseases, such as hypertension. The aim was to evaluate if iron biomarkers and vitamins in the blood are associated with blood pressure in European adolescents. Participants from the Healthy Lifestyle in Europe by Nutrition in Adolescence cross-sectional study (N = 1089; 12.5-17.5 y; 580 girls) were selected by complex sampling. Multilevel linear regression models examined the associations between iron biomarkers and vitamins in the blood and blood pressure; the analyses were stratified by sex and adjusted for contextual and individual potential confounders. A positive association was found in girls between RBC folate concentration and systolic blood pressure (SBP) (β = 3.19; 95% confidence interval [CI], 0.61-5.77), although no association between the vitamin serum biomarkers concentrations and diastolic blood pressure (DBP) was found. In boys, retinol was positively associated with DBP (β = 3.84; 95% CI, 0.51-7.17) and vitamin B6 was positively associated with SBP (β = 3.82; 95% CI, 1.46-6.18). In contrast, holotranscobalamin was inversely associated with SBP (β = -3.74; 95% CI, -7.28 to -0.21). Levels of RBC folate and vitamin B6 in blood may affect BP in adolescents. In this context, programs aimed at avoiding high BP levels should promote healthy eating behavior by focusing on the promotion of vegetable proteins and foods rich in vitamin B12 (i.e., white meat and eggs), which may help to achieve BP blood control in adolescents.
材料
货号
品牌
产品描述
Sigma-Aldrich
2-丙醇, suitable for HPLC, 99.9%
Sigma-Aldrich
2-丙醇, ACS reagent, ≥99.5%
Sigma-Aldrich
正己烷, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
正己烷, suitable for HPLC, ≥95%
Sigma-Aldrich
正己烷, ReagentPlus®, ≥99%
Sigma-Aldrich
2-丙醇, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
异丙醇, meets USP testing specifications
Sigma-Aldrich
视黄醇, synthetic, ≥95% (HPLC), (Powder or Powder with Lumps)
Sigma-Aldrich
2-丙醇, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
正己烷, Laboratory Reagent, ≥95%
Sigma-Aldrich
正己烷, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
2-丙醇, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
正己烷, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
异丙醇, ≥99.7%, FCC, FG
Sigma-Aldrich
2-丙醇, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
视黄醇, BioXtra, ≥97.5% (HPLC), ~3100 U/mg
Sigma-Aldrich
2-丙醇, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-丙醇, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
视黄醇, ≥95.0% (HPLC), ~2700 U/mg
Sigma-Aldrich
2-丙醇, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Sigma-Aldrich
2-丙醇, JIS special grade, ≥99.5%
Sigma-Aldrich
2-丙醇, suitable for HPLC, 99.5%
Sigma-Aldrich
2-丙醇, anhydrous, 99.5%
Sigma-Aldrich
2-丙醇, for molecular biology, BioReagent, ≥99.5%
Sigma-Aldrich
正己烷, JIS special grade, ≥96.0%
USP
2-丙醇, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
正己烷, anhydrous, 95%
Supelco
正己烷, analytical standard
Sigma-Aldrich
2-丙醇, SAJ first grade, ≥99.0%
Sigma-Aldrich
2-丙醇, ACS reagent, ≥99.5%