SU860145
1 of 10
Supelco
Z166065
橡胶塞
27232
瓶塞
Sigma-Aldrich
Z124591
Suba-Seal®隔膜
27235-U
27144
棕褐色 PTFE/硅胶隔垫
SYNF602015C
Synthware™ Buchner funnel
27018
带有 PTFE/硅胶隔垫的带孔螺纹盖
24886-U
隔垫型隔膜
27016
铝压接密封(无隔垫),银铝
854980-U
带 PTFE/橡胶隔垫的 11mm 钳口密封盖
gray butyl rubber
pkg of 1000 ea
20 mm
for use with 20 mm finish crmp vialsfor use with PerkinElmer headspace vials
正在寻找类似产品? 访问 产品对比指南
历史批次信息供参考:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.