Merck
57316-U
Supelco
SPME Fiber Assembly
greener alternative
for use with manual holder, Nitinol-core (NIT), 24 ga
登录查看公司和协议定价
About This Item
UNSPSC代码:
41115724
NACRES:
NB.21
产品名称
SPME 光纤组件未涂熔融石英, needle size 24 ga, for use with manual holder
物料
fused silica fiber
plain orange hub
质量水平
针尺寸
24 ga
环保替代产品特性
Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
技术
solid phase microextraction (SPME): suitable
光纤长度
1 cm
应用
food and beverages
相容性
for use with manual holder
环保替代产品分类
相关类别
一般描述
我们致力于为您提供绿色替代产品,这些产品遵守一项或多项绿色化学12项原则。这是一种固相微萃取(SPME)技术产品。SPME是一种更为环保的样品制备技术,因为它是一种无溶剂提取技术,SPME纤维头可以重复使用并且产生的废物极少,因此符合“防止产生废物”和“更安全的溶剂和助剂”原则。点击 此处 以获取更多信息。
历史批次信息供参考:
分析证书(COA)
访问文档库
相关内容
Solid phase microextraction (SPME) is a solvent-free sample preparation technique that uses coated fibers to extract analytes of interest from a sample before analysis by GC or HPLC.
固相微萃取(SPME)是一种无溶剂样品制备技术,利用涂层纤维头从样品中提取目标分析物,以便进行后续GC或HPLC分析。