27108
1 of 10
Supelco
29378-U
经过认证的螺纹口样品瓶套装,12×32mm,9mm 螺纹,未组装
29319-U
经过认证的带隔垫聚丙烯盖
27236
PTFE/硅胶隔垫
29403-U
经过认证的钳口样品瓶,12×32mm
29320-U
Z188700
Wheaton 样品瓶
507482
29127-U
12×32mm 广口钳口样品瓶便利套装
29372-U
经过认证的螺纹口样品瓶,9mm 螺纹,12×32mm
Z291633
12×32mm 标准螺纹自动进样器样品瓶
red PTFE/silicone/red PTFE septumsilver aluminum seal
closure type crimp seal
pkg of 100 ea
11 mm
thread for 11 mm
正在寻找类似产品? 访问 产品对比指南
Analytical Chemistry
Analytical Chromatography
Analytical Vials
历史批次信息供参考:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.