主要文件
Z329339
BRAND® magnetic stirring bar, PTFE, triangular
L 25 mm, bar diam. 8 mm
登录查看公司和协议定价
About This Item
UNSPSC代码:
41103806
eCl@ss:
32100100
NACRES:
NB.72
物料
triangular PTFE-coated
包装
pack of 10
制造商/商品名称
BRAND 137810
长度
25 mm
柱直径
8 mm
正在寻找类似产品? 访问 产品对比指南
一般描述
Wedge shaped. Ideal for dissolving solids and mixing of sediments due to scraping action. Angular surface creates high turbulence even at low speeds.
其他说明
BR137810 (India)
法律信息
BRAND is a registered trademark of BRAND GMBH + CO KG
历史批次信息供参考:
分析证书(COA)