Merck
BR122538
BRAND®实验室瓶子,有刻度和螺口盖
capacity 100 mL
别名:
实验室瓶子, 玻璃, 瓶子
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121800
NACRES:
NB.43
材料
borosilicate glass bottle
clear bottle
narrow-mouth bottle
polypropylene cap
round bottle
接合處
Joints threaded neck (GL45)
無菌
non-sterile
特點
autoclavable
graduations
with cap
包裝
pack of 10 ea
製造商/商標名
BRAND 122538
容量
100 mL
直徑× 高度
56 mm × 105 mm
正在寻找类似产品? 访问 产品对比指南
一般說明
Threaded to DIN ISO 4796. Studded base for stability and abrasion resistance. Graduated, with marking area. Screw cap and pouring ring PP. Autoclavable (121 °C).
法律資訊
BRAND is a registered trademark of BRAND GMBH + CO KG