主要文件
HS15907
Spatula with Scraper Round End & Tapered Flat End
stainless steel
登录查看公司和协议定价
About This Item
UNSPSC代码:
41122403
NACRES:
NB.71
物料
stainless steel
无菌性
non-sterile
包装
pack of 1 ea
制造商/商品名称
Heathrow Scientific HS15907
长度 × 宽度
195 mm × 7.9 mm
一般描述
Spatula transfers, lifts, and handles powders and granular materials.
特点和优势
  • Made from nickel-plated, stainless steel wire
  • Mirror finish with round flat and tapered flat ends
  • Autoclavable
历史批次信息供参考:
分析证书(COA)