主要文件
G2775
Sigma-Aldrich
Gaskets for vertical mini gel
10 cm wide, size 1mm x 8cm, color-coded red
登录查看公司和协议定价
About This Item
UNSPSC代码:
12352200
NACRES:
NB.51
产品名称
Gaskets for vertical mini gel 10 cm wide, size 1 mm × 8 cm, color-coded red
物料
silicone
质量水平
尺寸
1 mm × 8 cm
颜色
color-coded red
正在寻找类似产品? 访问 产品对比指南
特点和优势
Unique gasket provides continuous seal around sides and bottom of glass plates, eliminating the need for a bottom spacer. Gasket thickness must match comb and spacers. Length of gasket material is approx. 26 cm (2 × h + w). Gaskets are color-coded for easy identification.
历史批次信息供参考:
分析证书(COA)