主要文件
EP5820772000
Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges
F-35-48-17, to hold, 40 x 15 mL conical tubes, with 48 steel sleeves and adapters
登录查看公司和协议定价
About This Item
UNSPSC代码:
41103909
NACRES:
NB.82
特点
to hold 40 x 15 mL conical tubes
high-speed fixed angle rotor
with 48 steel sleeves and adapters
包装
pack of 1 ea
制造商/商品名称
Eppendorf® 5820772000 (Global)
参数
20,913 × g max. RCF
技术
centrifugation: suitable
相容性
for use with centrifuge models 5804/5804R/5810/5810R
正在寻找类似产品? 访问 产品对比指南
一般描述
A wide selections of rotors to fit the Eppendorf 5804 and 5810 centrifuges.
法律信息
Eppendorf is a registered trademark of Eppendorf AG
历史批次信息供参考:
分析证书(COA)