Merck
EP5820722002
Adapter for Eppendorf® FA-45-6-30 Rotor
capacity, 1 x 35 mL tube, pack of 2
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41103911
NACRES:
NB.82
特點
capacity 1 x 35 mL tube
for use with aerosol-tight lid
包裝
pack of 2
製造商/商標名
Eppendorf® 5820722002 (Global)
參數
16,161 × g max. RCF (at 11,000 rpm for 5804/5804 R/5810)
技術
centrifugation: suitable
直徑
29 mm
相容性
for use with FA-6x50
for use with Rotor FA-45-6-30
正在寻找类似产品? 访问 产品对比指南
一般說明
Adapters for various size tubes and bottles for use with Eppendorf rotor FA-45-6-30.
法律資訊
Eppendorf is a registered trademark of Eppendorf AG