跳转至内容
Merck
所有图片(1)

主要文件

D6929

Sigma-Aldrich

2,5-Diaziridinyl-1,4-benzoquinone

别名:

2,5-Bis(1-aziridinyl)-1,4-benzoquinone, DZQ

登录查看公司和协议定价


About This Item

经验公式(希尔记法):
C10H10N2O2
CAS号:
分子量:
190.20
MDL编号:
UNSPSC代码:
12352103

生化/生理作用

Bifunctional alkylating agent, inducing DNA interstrand cross-linking. Most efficient at low pH and following reductive conditions.

历史批次信息供参考:

分析证书(COA)

Lot/Batch Number

没有发现合适的版本?

如果您需要特殊版本,可通过批号或批次号查找具体证书。

已有该产品?

在文件库中查找您最近购买产品的文档。

访问文档库

M D Berardini et al.
Biochemistry, 32(13), 3306-3312 (1993-04-06)
The nucleotide sequence preferences for the formation of interstrand cross-links induced in DNA by 2,5-diaziridinyl-1,4-benzoquinone (DZQ) and 3,6-dimethyl-2,5-diaziridinyl-1,4-benzoquinone (MeDZQ) were studied using synthetic duplex oligonucleotides and denaturing polyacrylamide gel electrophoresis (PAGE). Reaction of these bifunctional alkylating agents with a DNA
J A Hartley et al.
Biochemistry, 30(50), 11719-11724 (1991-12-17)
Several bifunctional alkylating agents of the aziridinylbenzoquinone class have been evaluated as potential antitumor agents. 3,6-Bis[(2-hydroxyethyl)amino]-2,5- diaziridinyl-1,4-benzoquinone (BZQ), 2,5-diaziridinyl-1,4-benzoquinone (DZQ), 3,6-bis(carboxyamino)-2,5-diaziridinyl- 1,4-benzoquinone (AZQ), and six analogues of AZQ have been studied for their ability to induce DNA interstrand cross-linking, as
Alley, S.C., et al.
Journal of the American Chemical Society, 116, 2734-2734 (1994)

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系客户支持